Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
3,3',5,5'-Tetramethylbenzidine (TMB) substrate, ReadiUse™ (colorimetric)

3,3',5,5'-Tetramethylbenzidine (TMB) substrate, ReadiUse™ (colorimetric)

Plastic bottle, 1 L
Catalog # 76481-872
Irritant
Supplier:   AAT Bioquest
Supplier #:  11003
3,3',5,5'-Tetramethylbenzidine (TMB) substrate, ReadiUse™ (colorimetric)
Catalog # 76481-872
Supplier:  AAT Bioquest
CAS Number:  54827-17-7
3,3',5,5'-Tetramethylbenzidine (TMB) substrate, ReadiUse™ (colorimetric)
3,3',5,5'-Tetramethylbenzidine (TMB) substrate, ReadiUse™ (colorimetric)

Plastic bottle, 1 L

CAS Number:  54827-17-7
Price
$646.22
Log in for availability
Each (1l)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Horseradish peroxidase (HRP) and HRP conjugates facilitate the ABTS oxidation in the presence of hydrogen peroxide, turning ABTS into its blue-green oxidized product.

  • easy colorimetric detection
  • easily measured via spectrophotometer
Synonyms:  TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine
  • CAS Number:54827-17-7
  • Formula:C₁₆H₂₀N₂
  • MDL Number:MFCD00007748
  • Molecular Weight:240.35 g/mol
  • ADR:5,II

Documents

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    1 L

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.