$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
PIPETTE M12-50 TWELVE-CHANNEL 5-50 UL
MP76383-118
:
Brandtech
PIPETTE M12-50 TWELVE-CHANNEL 5-50 UL
MP76383-118
:
Brandtech
:
705926TU
Brandtech Pipette Trade-Up
BrandTech Pipette Trade-Up CA
BrandTech Pipette Trade-Up US
BrandTech Transferpette Pipette.pdf