Specifications
- Volume:5 - 50 µl
- For use with:2 - 200 µl tips
- Increments:0.1 µl
- Accuracy:±0.8%
- Precision:≤0.4%
Specifications
About this item
Transferpette® S pipettes are lightweight, ergonomic, fully autoclavable, and work with a wide range of tip brands.
True one-hand operation for right- and left-handed users; easily set the volume, pipette, and eject the tip
Color code on volume adjustment wheel, shaft, and volume-change protection
Low operating forces and a short stroke ensure relaxed pipetting, even during long pipetting series
Easy insertion and ejection of tips due to O-rings on multi-channel pipettes
Universal tip cone
Fully autoclavable at 121 °C
The volume-change protection is easy to operate with gloves on, and protects the set volume against accidental adjustment. The Transferpette® S also features Easy Calibration technology, which makes calibration a breeze. Calibration can be carried out without tools. A compatibility table shows which tips fit which device, so that the correct pipette tips can always be ordered.
Supplied with performance certificate, battery, power adapter, reagent reservoir, installation tool, and silicone grease.