Order Entry
Puerto Rico
ContactUsLinkComponent
PIPETTE ELECTRONIC 8-CHANNEL 100UL
BRAND® Transferpette® -8/-12 Multi-Channel Electronic Pipettes, Trade-Up Promotion
Catalog # MP12000-508
Supplier:  Brandtech
BRAND® Transferpette® -8/-12 Multi-Channel Electronic Pipettes, Trade-Up Promotion
Catalog # MP12000-508
Supplier:  Brandtech
Supplier Number:  705423TU

Specifications

  • Volume:
    5 - 100 µl
  • For use with:
    2 - 200 µl tips
  • Increments:
    0.1 µl
  • Accuracy:
    ±0.8%
  • Precision:
    ≤0.25%

Specifications

About this item

Transferpette® electronic pipettes are lightweight, ergonomic, fully autoclavable, and work with a wide range of tip brands.

  • True one-hand operation for right- and left-handed users; easily set the volume, pipette, and eject the tip

  • Color code on volume adjustment wheel, shaft, and volume-change protection
  • Low operating forces and a short stroke ensure relaxed pipetting, even during long pipetting series
  • Easy insertion and ejection of tips due to O-rings on multi-channel pipettes
  • Universal tip cone
  • Fully autoclavable at 121 °C

The volume-change protection is easy to operate with gloves on, and protects the set volume against accidental adjustment. The Transferpette® electronic also features easy calibration technology, which makes calibration a breeze. Calibration can be carried out without tools. A compatibility table shows which tips fit which device, so that the correct pipette tips can always be ordered.

Delivery:

Supplied with performance certificate, battery, power adapter, reagent reservoir, installation tool, and silicone grease.