$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
KIT DETECTION HORSE RAB IGG POLYMER 15ML
Catalog # 103881-252
Supplier:
Vector Laboratories
Expand image
KIT DETECTION HORSE RAB IGG POLYMER 15ML
Catalog # 103881-252
Supplier:
Vector Laboratories
Supplier Number:
MP-6401-15