Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
KIT DETECTIN HORSE GOAT IGG POLYMER 15ML
Catalog # 103881-246
Supplier:
Vector Laboratories
Expand image
KIT DETECTIN HORSE GOAT IGG POLYMER 15ML
Catalog # 103881-246
Supplier:
Vector Laboratories
Supplier Number:
MP-5405-15