Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
BUFFER CATALYST 10X F/LIGATION 1500UL
Catalog # 103881-098
Supplier:
Vector Laboratories
Expand image
BUFFER CATALYST 10X F/LIGATION 1500UL
Catalog # 103881-098
Supplier:
Vector Laboratories
Supplier Number:
S-2006-105