$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
RECOMBINANT NUCLEOSOME HU H3K27ME3 20 UG
MSPP-16-0317
:
EPICYPHER INC MS
RECOMBINANT NUCLEOSOME HU H3K27ME3 20 UG
MSPP-16-0317
:
EPICYPHER INC MS
:
16-0317