CELLS PERIPHERAL BLOOD B HU 1X10E7 CELLS
Catalog # MSPP-70023
Supplier: Stemcell Technologies
CELLS PERIPHERAL BLOOD B HU 1X10E7 CELLS
Catalog # MSPP-70023
Supplier: Stemcell Technologies
Supplier Number: 70023
Frequently Bought Together
VWR® VividPro™ Fluorescent Stain
Supplier: VWR
KIT POWER SNAP ELECTROPHORSIS EX1% 240V
Supplier: Invitrogen
ANTIBODY R840 HLA-DR L243 HUMAN 25TEST
Supplier: CYTEK BIOSCIENCES, INC. MS
Blot Boxes, IBI Scientific
Supplier: IBI Scientific
OSM (227AA) HUMAN >95% 50UG
Supplier: Genscript
Tris Buffers, Sterile
Supplier: G-Biosciences
IPG Buffers, Cytiva
Supplier: Cytiva
Sapphire Prestained Protein Standard 10 to 180 kDa, Rockland Immunochemicals
Supplier: Rockland Immunochemical
Buffer P1 (Red), Zymo Research
Supplier: Zymo Research
2-D Quant Kit, Cytiva
Supplier: Cytiva
Antibody Fab Fragmentation, G-Biosciences
Supplier: G-Biosciences
RNA RECOVERY BUFFER 10 ML
Supplier: Zymo Research
Zyppy Elution Buffer, Zymo Research
Supplier: Zymo Research
1X PBS Buffer with 1% Casein
Supplier: Rockland Immunochemical
FGF-BASIC (154AA) HUMAN >95% 50UG
Supplier: Genscript
GelBond® Film
Supplier: Lonza
TrueBlot® IP/Western Blot, Rockland Immunochemicals
Supplier: Rockland Immunochemical