Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
RECOMBINANT SREBF1 PROTEIN ECOLI 50 UG
Catalog # MSPP-MBS203320
Supplier:
MYBIOSOURCE
Expand image
RECOMBINANT SREBF1 PROTEIN ECOLI 50 UG
Catalog # MSPP-MBS203320
Supplier:
MYBIOSOURCE
Supplier Number:
MBS203320750UG