Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Raised Panel Polyester Rectangular Labels, 3" Core, Brady

Raised panel labels, Polyester, White, 3", 25.4×50.8 mm (1×2"), 500 labels/roll
  76461-154
 :  THTEP-173-593-.5
Raised Panel Polyester Rectangular Labels, 3" Core, Brady
  76461-154
Raised Panel Polyester Rectangular Labels, 3" Core, Brady
Raised Panel Polyester Rectangular Labels, 3" Core, Brady

Raised panel labels, Polyester, White, 3", 25.4×50.8 mm (1×2"), 500 labels/roll

 
$397.28
   
Roll of 1
 

Raised panel Adhesive-Taped Polyester (B-593) labels are designed for patch panel identification, external push-buttons, switches, and internal connection points.

  • Raised panel design
  • Aggressive adhesive
  • Solvent-resistant

Raised panel Adhesive-Taped Polyester (B-593) labels can be used as rating and serial plates using alphanumerics that require nameplate quality. Aggressive adhesive and solvent resistance makes them a great replacement for engraved tags. Service temperature of ‒20 to +100 °C (‒4 to +212 °F)

Labels are designed for use with BBP®81 Label Printer, Brady IP® Series Label Printer, BradyPrinter i5100 Industrial Label Printer, BradyPrinter i7100 Industrial Label Printer, THT 3" Core Printers.

 : UL approved.

 : A ribbon is required to print this material. Use Part Number 49006-110 for the BradyPrinter i5100 or BRADY IP® Series Label Printer; use R6006 for a different printer.

 
  • Color:
    White
  • Description:
    Raised panel labels
  • Dimensions:
    25.4×50.8 mm (1×2")
  • Material:
    Polyester
  • Packaging:
    500 labels/roll
  • Core size:
    3"