ChloraPrep® Applicators with Sterile Solution, BD Diagnostics
A comprehensive skin antiseptic applicator collection, available in six sizes and three tints.
- The only 2% CHG/70% IPA solution with >50 clinical studies supporting its efficacy and safety
- A wide range of coverage areas to fit varied clinical needs, allowing for cost-saving standardized skin prep across care settings
ChloraPrep™ Applicators with Sterile Solution are used for rapid-acting and persistent preoperative skin preparation. Its proven formulation is available in six unique applicators designed for procedures ranging from peripheral IV insertion to major surgery. The applicator allows for an easy application of the unique formulation for skin preparation, available in 1 ml, Frepp™ 1.5 ml, 3 ml, 10.5 ml and 26 ml sizes. Designed to fit the patient's skin tone, use Hi-Lite Orange for lighter pigmented skin, Scrub Teal for darker pigmented skin, or clear for a colorless effect.
- Color:Clear
- Description:ChloraPrep® applicator with sterile solution
- Size:1 ml