Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
ISOTYPE CONTROL MS C1.18.4 IGG2A 100UG
Catalog # 76447-270
Supplier:
Tonbo Biosciences
Expand image
ISOTYPE CONTROL MS C1.18.4 IGG2A 100UG
Catalog # 76447-270
Supplier:
Tonbo Biosciences
Supplier Number:
85-4724-U100
Related Information
Documentation
Product Literature
Tonbo Biosciences violetFluor™ Antibodies
Documentation