Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
ISOTYPE CONTROL RAT LTF-2 IGG 25UG
Catalog # 76447-232
Supplier:
Tonbo Biosciences
Expand image
ISOTYPE CONTROL RAT LTF-2 IGG 25UG
Catalog # 76447-232
Supplier:
Tonbo Biosciences
Supplier Number:
85-4031-U025
Related Information
Documentation
Product Literature
Tonbo Biosciences violetFluor™ Antibodies
Documentation