$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
RECOMBINANT ANTI-AAV2 A20 MOUSE AB 50 UG
MSPP-610298
:
Cedar Lane
RECOMBINANT ANTI-AAV2 A20 MOUSE AB 50 UG
MSPP-610298
:
Cedar Lane
:
610298