mPrep/f™ Pipette Filter Couplers, Electron Microscopy Sciences
Product Details & Documents
Filter couplers prevent the introduction of damaging reagents into pipettors. They also improve the fit of mPrep/g™ capsules on some pipettors.
Available in two pore sizes. Standard mPrep/f30™ has a nominal 30 µm pore size filter appropriate for most applications. Extreme mPrep/f13™ has a nominal 13 µm pore size filter for use with biohazards and very aggressive reagents.
Delivery information: Pack includes 16 filter couplers and a capsule storage box.
Specifications
- Tip style:Filter-couplers, f30 - Standard