Orders LinkContactUsLinkComponent
mPrep™ Pipettor Kits, Electron Microscopy Sciences
mPrep™ Pipettor Kits, Electron Microscopy Sciences
Single channel pipettor, one mPrep™ capsule pack of your choice and one pack of 96 pipette tips (size 10 µl), 200 µl
Catalog # 76439-626
Supplier #:  85010-07
mPrep™ Pipettor Kits, Electron Microscopy Sciences
Catalog # 76439-626
mPrep™ Pipettor Kits, Electron Microscopy Sciences
mPrep™ Pipettor Kits, Electron Microscopy Sciences
Single channel pipettor, one mPrep™ capsule pack of your choice and one pack of 96 pipette tips (size 10 µl), 200 µl
Price
$1,053.85
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Choose from either single- or multi-channel pipettor kits built around mPrep/g™ capsules.
Single-channel kit includes single channel 200 µl pipettor, one mPrep™ capsule pack of your choice and one pack of 96 pipette tips, size 10 µl.
Eight-channel kit includes eight-channel 200 µl pipettor, one mPrep™ capsule pack of your choice and one pack of 96 pipette tips, size 10 µl.
Specifications
  • Volume:
    200 µl
  • Description:
    Single channel pipettor, one mPrep™ capsule pack of your choice and one pack of 96 pipette tips (size 10 µl)