Orders LinkContactUsLinkComponent
Silicon Dioxide TEM Window Grids, Electron Microscopy Sciences
Silicon Dioxide TEM Window Grids, Electron Microscopy Sciences
1000 µm, 100 nm
Catalog # 76439-612
Supplier #:  76042-92
Silicon Dioxide TEM Window Grids, Electron Microscopy Sciences
Catalog # 76439-612
Silicon Dioxide TEM Window Grids, Electron Microscopy Sciences
Silicon Dioxide TEM Window Grids, Electron Microscopy Sciences
1000 µm, 100 nm
Price
$522.05
Log in for availability
Pack of 10
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
By making the grids slightly narrower users now have easy access to grids in TEM holders. No more fumbling with tweezers while trying to pick up or put down grids. The new TEM Window grid shape is still compatible with all standard holders. TEM Window dimensions are 2.9 mm in diameter and 100 µm thick.
  • Plasma cleanable - can be vigorously plasma cleaned to remove organic contamination
  • Field to field uniformity - reduced variability
  • Tolerates temperatures above 1000 °C - supports use in environmental TEMs where dynamic processes are observed at high temperatures
  • Withstands harsh conditions - Provides an ideal balance of imaging resolution, chemical stability and mechanical strength
  • Incorporates stoichiometric silicon dioxide - offers the ability to analyze for nitrogen by EDX techniques
Specifications: 100 µm thick frame, fits 3 mm sample holders.
Non-porous films are lightly wrinkled with approximately 5 µm or less deflection across 100 µm of travel. This is typically not problematic for high-resolution imaging.
GFLAT silicon oxide films are created by a proprietary process that uniquely results in flat, suspended silicon oxide membranes. These membranes are ideally suited for biological imaging studies, with a glass-like hydrophilic surface. These TEM Windows are essentially micro-scale glass cover slips.
Specifications
  • Dimensions:
    1000 µm
  • Thickness:
    100 nm