Orders LinkContactUsLinkComponent
Pure Silicon TEM Windows, Electron Microscopy Sciences
Pure Silicon TEM Windows, Electron Microscopy Sciences
(8) 100 µm, (1) 100×350 µm, 9 nm
Catalog # 76439-596
Supplier #:  76042-74
Pure Silicon TEM Windows, Electron Microscopy Sciences
Catalog # 76439-596
Pure Silicon TEM Windows, Electron Microscopy Sciences
Pure Silicon TEM Windows, Electron Microscopy Sciences
(8) 100 µm, (1) 100×350 µm, 9 nm
Price
$1,013.23
Log in for availability
Pack of 10
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Pure silicon TEM windows feature imaging windows with 5 to 35 nm thickness, reducing background contribution and interference for higher contrast imaging. Most impressively, 5 nm thick non-porous pure silicon TEM Windows are thinner than the thinnest commercially available amorphous carbon membranes.
  • Plasma cleanable - can be vigorously plasma cleaned to remove organic contamination, unlike traditional carbon grids
  • Silicon composition - Sputter-deposited, pure, intrinsic silicon
  • Minimal background signal - Enables elemental analyses of samples containing nitrogen and/or carbon
  • Hydrophilicity - The hydrophilicity of both non-porous and porous Pure Silicon TEM Windows is tunable by plasma and/or ozone treatment making sample preparation easier, particularly for samples in aqueous solutions
  • Increased stability - At high beam currents and high annealing temperatures (600 °C for non-porous, >1000 °C for nanoporous)
Field to field uniformity - Non-porous pure silicon TEM Windows are more consistently thin than carbon grids, reducing field-to-field variability. (Note: Porous windows do have inherent crystalline features, but feature background-free nanometer-scale pores).
Reduced chromatic blur - In comparison to the thinnest commercially available amorphous carbon membranes, 5 nm Non-porous pure silicon TEM Windows yield half the chromatic blur. This dramatic difference results from a two-fold reduction in inelastic scattering of electrons passing through the thinner membranes of silicon TEM Windows. In turn, the reduced chromatic blur offers a potential two-fold improvement in imaging resolution.
Nanometer-scale pores - Pure Silicon TEM Windows are available as porous films with pores ranging from 5 to 50 nm in diameter. The pores allow simple and stable suspension of nanoscale materials for imaging without intervening background.
Isolated poly-crystallinity - The poly-crystalline nature of porous pure silicon TEM windows offers an internal calibration standard for x-ray diffraction studies. The isolated crystalline features also provides a convenient and reliable scale for high-resolution size measurements, well-characterized crystal lattice of silicon.
Nanoporous - Using P30 membranes has made the nanoporous TEM windows significantly more porous. Pore sizes have increased to include a range of pores from 10 to 60 nm in diameter. Non-porous films are lightly wrinkled with approximately 5 µm or less deflection across 100 µm of travel. This is typically not problematic for high-resolution imaging.
Specifications
  • Dimensions:
    (8) 100 µm, (1) 100×350 µm
  • Thickness:
    9 nm