Orders LinkContactUsLinkComponent
EMS Grid Prep Holders, Electron Microscopy Sciences
EMS Grid Prep Holders, Electron Microscopy Sciences
Grid prep holder, capacity 36, thickness 6 mm, 43×43 mm
Catalog # 76439-522
Supplier #:  71175-01
EMS Grid Prep Holders, Electron Microscopy Sciences
Catalog # 76439-522
EMS Grid Prep Holders, Electron Microscopy Sciences
EMS Grid Prep Holders, Electron Microscopy Sciences
Grid prep holder, capacity 36, thickness 6 mm, 43×43 mm
Price
$192.65
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
EMS preparation tray may be use for staining and/or placing an evaporator for the coating of grids.
  • Medical grade aluminum
  • Includes a plastic storage box
These unique trays have a M4 tapped hole in the center which allows for insertion of pin mount. They also have a screw pin in the middle that may be used as a handle for manipulating the trays.
Specifications
  • Description:
    Grid prep holder, capacity 36, thickness 6 mm
  • Dimensions:
    43×43 mm