Picking Trays, Electron Microscopy Sciences
Product Details & Documents
Trays perfect for placement of grids and storage of small components. The trays feature 42 cells with a lip differentiating each cell. The cells measure 11×11 mm (⁷/₁₆×⁷/₁₆").
These unique trays are available in either black rigid ABS plastic or aluminum in black or tan.
Specifications
- Description:Picking tray, Aluminum, black
- Dimensions:96×34 mm (3.78×1.34")