Orders LinkContactUsLinkComponent
Picking Trays, Electron Microscopy Sciences
Picking tray, ABS plastic, black, 96×34 mm (3.78×1.34")
  76439-514
 :  71171-01
Picking Trays, Electron Microscopy Sciences
  76439-514
Picking Trays, Electron Microscopy Sciences
Picking Trays, Electron Microscopy Sciences
Picking tray, ABS plastic, black, 96×34 mm (3.78×1.34")
$39.08
Each
Trays perfect for placement of grids and storage of small components. The trays feature 42 cells with a lip differentiating each cell. The cells measure 11×11 mm (⁷/₁₆×⁷/₁₆").
These unique trays are available in either black rigid ABS plastic or aluminum in black or tan.
  • Description:
    Picking tray, ABS plastic, black
  • Dimensions:
    96×34 mm (3.78×1.34")