Orders LinkContactUsLinkComponent
Horizontal Grid Boxes, Electron Microscopy Sciences
Horizontal Grid Boxes, Electron Microscopy Sciences
Horizontal grid box
Catalog # 76439-512
Supplier #:  71161-03
Horizontal Grid Boxes, Electron Microscopy Sciences
Catalog # 76439-512
Horizontal Grid Boxes, Electron Microscopy Sciences
Horizontal Grid Boxes, Electron Microscopy Sciences
Horizontal grid box
Price
$403.02
Log in for availability
Pack of 8
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These slide-in type grid boxes offer easy handling of the grids without damaging the support film.
Specifications
  • Description:
    Horizontal grid box