Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

COD Standards, Environmental Express®

COD standard, 250 ml, 10000 mg/L
Catalog # 76436-810
 :  B1031
COD Standards, Environmental Express®
Catalog # 76436-810
CAS Number:  
COD Standards, Environmental Express®
COD Standards, Environmental Express®

COD standard, 250 ml, 10000 mg/L

CAS Number:  
Price
$46.54
Log in for availability
Each (250ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Controls approved for use in both EPA-compliant and mercury-free vials.

  • Made from primary standard grade potassium hydrogen phthalate
  • Delivered with an SDS and minimum 8 month shelf life (refrigeration required)
  • Easy-to-follow dilution directions printed directly on the label

 : This product complies with the following methods: EPA 410.4, SM 5220D-2011, HACH 8000, ASTM D1252-06, ISO 15705:2002, USGS I-3561-06, BS 6068-2.34:1988, DIN 38414-9, and ISO 6060:1989.

 

Specifications

Specifications

  • Concentration:
    10000 mg/L
  • Description:
    COD standard
  • Size:
    250 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.