Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

COD Reagent Vials, Environmental Express®

COD reagent tubes Hg-free, Vial 16 x 100 mm, 0 - 150 mg/L, 100/pack
Catalog # 76436-796
 :  B0010
COD Reagent Vials, Environmental Express®
Catalog # 76436-796
CAS Number:  
COD Reagent Vials, Environmental Express®
COD Reagent Vials, Environmental Express®

COD reagent tubes Hg-free, Vial 16 x 100 mm, 0 - 150 mg/L, 100/pack

CAS Number:  
Price
$237.63
Log in for availability
Pack of 1
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Obtain consistent and accurate results while saving money.

  • Pre-filled reagent vials for either EPA-approved or mercury-free COD analysis
  • Quality analyzed to provide you hassle-free consistent results
  • Don''t change your instrument or method—vials work with Hach, Orbeco, Chemetrics, and Thermo COD systems

Pre-filled with COD reagents, these digestion vials come in either EPA-approved or mercury-free versions for low-, standard-, and ultra-high-range analysis. Each lot is tested to ensure superior quality from the cap and vial to the chemical contents. Glass vials are 16 mm x 100 mm to work with your existing COD reactor and spectrophotometer. Simply add 2 mL of your sample, heat in your digestor, and read in your spectrometer.

Although mercury-free vials are not approved for EPA Methods, they may be used for non-EPA COD testing when chlorine interferrence is not a factor. Ultra-high-range reagent vials have also not approved for EPA testing methods. COD digestion vials may contain acid, mercury, and silver and must be disposed of properly.

 : Supplied with C of A and SDS.

Specifications

Specifications

  • Description:
    COD reagent tubes Hg-free
  • Size:
    Vial 16 x 100 mm
  • Range:
    0 - 150 mg/L
  • No. of tests:
    100/pack

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.