Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
igScript™ Probe-Based One Step RT-qPCR Kit, Intact Genomics

igScript™ Probe-Based One Step RT-qPCR Kit, Intact Genomics

igScript™ Probe-Based One Step RT-qPCR Kit, 5000
Catalog # 103853-762
Marketsource
Supplier:   Intact Genomics
Supplier #:  4219
igScript™ Probe-Based One Step RT-qPCR Kit, Intact Genomics
Catalog # 103853-762
Supplier:  Intact Genomics
Marketsource
CAS Number:  
igScript™ Probe-Based One Step RT-qPCR Kit, Intact Genomics

igScript™ Probe-Based One Step RT-qPCR Kit, 5000

CAS Number:  
Price
$3,750.00
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

igScript™ Probe-Based One Step RT-qPCR Kit provides improved PCR efficiency, wider dynamic range, superior sensitivity and specificity.

  • Enhanced efficiency, specificity, and sensitivity
  • Compatible with all real-time PCR instruments
  • Superior gene expression results under various cycling conditions
  • Robust and active for cDNA synthesis at temperatures up to 55 °C

IgScript™ Probe-Based One Step RT-qPCR 2× master mix contains IgScript™ Reverse Transcriptase, Taq DNA polymerase, MgCl₂, dNTPs, stabilizers and low ROX reference dye with standard buffer providing improved RT-qPCR efficiency, wider dynamic range, superior sensitivity and specificity. IgScript™ Probe-Based One Step RT-qPCR 2× master mix is a ready-to-use cocktail containing all components except primers, probe and template, for the amplification and detection of DNA in RT-qPCR. This 2× master mix requires minimal handling during reaction setup and offer consistent and robust RT-qPCR reactions.

IgScript™ Reverse Transcriptase is a recombinant MMLV reverse transcriptase with reduced RNase H activity, increased thermostability and can produce cDNA from small amount of total RNA for real-time RT-qPCR analysis and other applications. Taq DNA Polymerase is a thermostable DNA polymerase that possesses a 5´→3´ polymerase (1, 2) and a 5´→3´ exonuclease activity (3, 4). The amplification step features a high quality Taq DNA Polymerase which offers robust, reliable and better amplification.

Delivery information: All our products are shipped with dry ice via FedEx overnight delivery. US orders received by 1:00 pm (Central time, Monday-Thursday) will typically be shipped on the same day in the US. Orders after 1:00 pm will be shipped on the next business day. Orders received on Friday will be ship on the following Monday.
The shipping and dry ice packaging cost is $40 for your entire order in the 48 contiguous United States.

Packaging: Product includes: IgScript™ Probe-Based One Step RT-qPCR 2× master mix.

Specifications

Specifications

  • Description:
    igScript™ Probe-Based One Step RT-qPCR Kit
  • No. of reactions:
    5000
  • Cat. No.:
    103853-762
  • One step:
    Yes
  • Application:
    RT-qPCR probe

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.