Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

igScript™ Probe-Based One Step RT-qPCR Kit, Intact Genomics

igScript™ Probe-Based One Step RT-qPCR Kit, 500
Catalog # 103853-758
Marketsource
Supplier:   Intact Genomics
 :  4215
igScript™ Probe-Based One Step RT-qPCR Kit, Intact Genomics
Catalog # 103853-758
Supplier:  Intact Genomics
Marketsource
CAS Number:  
igScript™ Probe-Based One Step RT-qPCR Kit, Intact Genomics

igScript™ Probe-Based One Step RT-qPCR Kit, 500

CAS Number:  
Price
$505.38
Log in for availability
Each
 
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

igScript™ Probe-Based One Step RT-qPCR Kit provides improved PCR efficiency, wider dynamic range, superior sensitivity and specificity.

  • Enhanced efficiency, specificity, and sensitivity
  • Compatible with all real-time PCR instruments
  • Superior gene expression results under various cycling conditions
  • Robust and active for cDNA synthesis at temperatures up to 55 °C

IgScript™ Probe-Based One Step RT-qPCR 2× master mix contains IgScript™ Reverse Transcriptase, Taq DNA polymerase, MgCl₂, dNTPs, stabilizers and low ROX reference dye with standard buffer providing improved RT-qPCR efficiency, wider dynamic range, superior sensitivity and specificity. IgScript™ Probe-Based One Step RT-qPCR 2× master mix is a ready-to-use cocktail containing all components except primers, probe and template, for the amplification and detection of DNA in RT-qPCR. This 2× master mix requires minimal handling during reaction setup and offer consistent and robust RT-qPCR reactions.

IgScript™ Reverse Transcriptase is a recombinant MMLV reverse transcriptase with reduced RNase H activity, increased thermostability and can produce cDNA from small amount of total RNA for real-time RT-qPCR analysis and other applications. Taq DNA Polymerase is a thermostable DNA polymerase that possesses a 5´→3´ polymerase (1, 2) and a 5´→3´ exonuclease activity (3, 4). The amplification step features a high quality Taq DNA Polymerase which offers robust, reliable and better amplification.

Delivery information: All our products are shipped with dry ice via FedEx overnight delivery. US orders received by 1:00 pm (Central time, Monday-Thursday) will typically be shipped on the same day in the US. Orders after 1:00 pm will be shipped on the next business day. Orders received on Friday will be ship on the following Monday.
The shipping and dry ice packaging cost is $40 for your entire order in the 48 contiguous United States.

Packaging: Product includes: IgScript™ Probe-Based One Step RT-qPCR 2× master mix.

Specifications

Specifications

  • Description:
    igScript™ Probe-Based One Step RT-qPCR Kit
  • No. of reactions:
    500
  • Cat. No.:
    103853-758
  • One step:
    Yes
  • Application:
    RT-qPCR probe

Specifications