Disposable Face Shields
Product 76417-632 is similar to 76423-496 but manufacturer and specifications may vary.
Product Details & Documents
Face shields are personal protective equipment (PPE) that are used for the protection of the facial areas (eyes, nose, mouth) from sprays, aerosols, and splashes.
- Latex free
- Made in an FDA registered facility
- Made from PETG
- Provides clarity of view
- Visors are anti-fogging
- Once size fits most
- Includes a comfortable forehead cushion
- Protective film on the shield (must be removed before use)
Ideal for healthcare and laboratory workers. Well suited for protection against COVID-19 and other pathogens.
Industries: Breweries, wineries and distilleries, Cannabis QA testing, clinical, cosmetics, environmental, food safety, compounding pharmacy, pharmaceutical manufacturing.
Documents
Pharma Microbiology Veterinary Microbiology
Additional Product Details
Specifications
- Color:Clear
- Description:Face shield, full face, disposable, anti-fogging plastic
- Material:PETG