Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Tyramide Amplification Buffer Plus, Biotium

Tyramide Amplification Buffer Plus, Biotium

Tyramide amplification buffer plus, 250
Catalog # 76423-466
Supplier:   Biotium
Supplier #:  22029-T
Tyramide Amplification Buffer Plus, Biotium
Catalog # 76423-466
Supplier:  Biotium
CAS Number:  
Tyramide Amplification Buffer Plus, Biotium
Tyramide Amplification Buffer Plus, Biotium

Tyramide amplification buffer plus, 250

CAS Number:  
Price
$145.20
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

An optimized buffer with enhanced sensitivity for tyramide signal amplification of immunostaining in cells or tissue sections.

  • For cell and tissue staining with tyramide signal amplification
  • Significantly improved brightness, specificity, and sensitivity
  • Compatible with CF® Dye and other tyramides
  • 250 or 1000 reaction sizes
  • 1× Tyramide amplification buffer plus
  • 30% Hydrogen peroxide

Tyramide amplification buffer plus is an optimized buffer with enhanced sensitivity for tyramide signal amplification of immunostaining in cells or tissue sections. It provides significantly improved brightness, specificity, and sensitivity compared to Biotium's original ready-to-use tyramide amplification buffer (76403-880, 76403-882).

Tyramide amplification buffer plus was developed as a new and improved version of Biotium's original ready-to-use tyramide amplification buffer (76403-880, 76403-882). It is also a component in all of Biotium's tyramide amplification kits.

Specifications

Specifications

  • Description:
    Tyramide amplification buffer plus
  • No. of reactions:
    250

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.