Specimen Stages, Electron Microscopy Sciences
Product Details & Documents
EMS Specimen Stages that meets most requirements.
These specimen stages are all easy-change, drop-in style with no scews, and are height adjustable, except for the rotary planetary stage.
Specifications
- Description:Combination of 76421-756 and small gearbox to offset sample position; Enable even coating up to a 4" sample size
- Stage:4" wafer and offset gearbox
- For use with:Q150R plus, Q150T