Orders LinkContactUsLinkComponent
Specimen Stages, Electron Microscopy Sciences
Specimen Stages, Electron Microscopy Sciences
6" wafer, Q300, Adjusts to accept 4 and 6" wafers comes with 76421-764, a 6" stub holder to accept up to 27¹/₈" pin stubs
  76421-758
Supplier #:  4500-4
Specimen Stages, Electron Microscopy Sciences
  76421-758
Specimen Stages, Electron Microscopy Sciences
Specimen Stages, Electron Microscopy Sciences
6" wafer, Q300, Adjusts to accept 4 and 6" wafers comes with 76421-764, a 6" stub holder to accept up to 27¹/₈" pin stubs
$2,260.67
Each
EMS Specimen Stages that meets most requirements.
These specimen stages are all easy-change, drop-in style with no scews, and are height adjustable, except for the rotary planetary stage.
  • Description:
    Adjusts to accept 4 and 6" wafers comes with 76421-764, a 6" stub holder to accept up to 27¹/₈" pin stubs
  • Stage:
    6" wafer
  • For use with:
    Q300