Orders LinkContactUsLinkComponent
Specimen Stages, Electron Microscopy Sciences
Specimen Stages, Electron Microscopy Sciences
4" wafer, All series, Adjusts to accept 2, 3 and 4" wafers comes with 76421-762, a 4" stub holder to accept up to 18¹/₈" pin stubs
Catalog # 76421-756
Supplier #:  4500-3
Specimen Stages, Electron Microscopy Sciences
Catalog # 76421-756
Specimen Stages, Electron Microscopy Sciences
Specimen Stages, Electron Microscopy Sciences
4" wafer, All series, Adjusts to accept 2, 3 and 4" wafers comes with 76421-762, a 4" stub holder to accept up to 18¹/₈" pin stubs
Price
$2,152.22
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
EMS Specimen Stages that meets most requirements.
These specimen stages are all easy-change, drop-in style with no scews, and are height adjustable, except for the rotary planetary stage.
Specifications
  • Description:
    Adjusts to accept 2, 3 and 4" wafers comes with 76421-762, a 4" stub holder to accept up to 18¹/₈" pin stubs
  • Stage:
    4" wafer
  • For use with:
    All series