Orders LinkContactUsLinkComponent
Specimen Stages, Electron Microscopy Sciences
Specimen Stages, Electron Microscopy Sciences
Slide stage, All series, Microscope slide stage for up to two 75×25 mm slides or eight stub positions for pin stubs
Catalog # 76421-752
Supplier #:  4500-17
Specimen Stages, Electron Microscopy Sciences
Catalog # 76421-752
Specimen Stages, Electron Microscopy Sciences
Specimen Stages, Electron Microscopy Sciences
Slide stage, All series, Microscope slide stage for up to two 75×25 mm slides or eight stub positions for pin stubs
Price
$1,765.42
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
EMS Specimen Stages that meets most requirements.
These specimen stages are all easy-change, drop-in style with no scews, and are height adjustable, except for the rotary planetary stage.
Specifications
  • Description:
    Microscope slide stage for up to two 75×25 mm slides or eight stub positions for pin stubs
  • Stage:
    Slide stage
  • For use with:
    All series