Specimen Stages, Electron Microscopy Sciences
Product Details & Documents
EMS Specimen Stages that meets most requirements.
These specimen stages are all easy-change, drop-in style with no scews, and are height adjustable, except for the rotary planetary stage.
Specifications
- Description:4-Place 25 mm stub stage with locking screws; May be fitted to 76421-742 rota cota stage
- Stage:4 place 25 mm stub
- For use with:All series