Orders LinkContactUsLinkComponent
Specimen Stages, Electron Microscopy Sciences
Specimen Stages, Electron Microscopy Sciences
14 place stub, All series, 14 places for 25 or 30 mm polished embedded samples; Includes a polished brass tally
Catalog # 76421-746
Supplier #:  4500-14
Specimen Stages, Electron Microscopy Sciences
Catalog # 76421-746
Specimen Stages, Electron Microscopy Sciences
Specimen Stages, Electron Microscopy Sciences
14 place stub, All series, 14 places for 25 or 30 mm polished embedded samples; Includes a polished brass tally
Price
$3,554.59
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
EMS Specimen Stages that meets most requirements.
These specimen stages are all easy-change, drop-in style with no scews, and are height adjustable, except for the rotary planetary stage.
Specifications
  • Description:
    14 places for 25 or 30 mm polished embedded samples; Includes a polished brass tally
  • Stage:
    14 place stub
  • For use with:
    All series