Orders LinkContactUsLinkComponent
Specimen Stages, Electron Microscopy Sciences
Specimen Stages, Electron Microscopy Sciences
6" square wafer, Q300T T plus, Stage to accept 6" square wafer or masks
Catalog # 76421-740
Supplier #:  4500-11
Specimen Stages, Electron Microscopy Sciences
Catalog # 76421-740
Specimen Stages, Electron Microscopy Sciences
Specimen Stages, Electron Microscopy Sciences
6" square wafer, Q300T T plus, Stage to accept 6" square wafer or masks
Price
$7,433.13
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
EMS Specimen Stages that meets most requirements.
These specimen stages are all easy-change, drop-in style with no scews, and are height adjustable, except for the rotary planetary stage.
Specifications
  • Description:
    Stage to accept 6" square wafer or masks
  • Stage:
    6" square wafer
  • For use with:
    Q300T T plus