Orders LinkContactUsLinkComponent
Specimen Stages, Electron Microscopy Sciences
Specimen Stages, Electron Microscopy Sciences
50 mm, All series, Standard stage with six stub positions for 15 mm or 6.5 mm or ¹/₈" pin stubs
Catalog # 76421-736
Supplier #:  4500-1
Specimen Stages, Electron Microscopy Sciences
Catalog # 76421-736
Specimen Stages, Electron Microscopy Sciences
Specimen Stages, Electron Microscopy Sciences
50 mm, All series, Standard stage with six stub positions for 15 mm or 6.5 mm or ¹/₈" pin stubs
Price
$695.98
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
EMS Specimen Stages that meets most requirements.
These specimen stages are all easy-change, drop-in style with no scews, and are height adjustable, except for the rotary planetary stage.
Specifications
  • Description:
    Standard stage with six stub positions for 15 mm or 6.5 mm or ¹/₈" pin stubs
  • Stage:
    50 mm
  • For use with:
    All series