Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
L(+)-Ascorbic acid 98%

L(+)-Ascorbic acid 98%

Catalog # 103846-132
Irritant
Marketsource
L(+)-Ascorbic acid 98%
Catalog # 103846-132
Supplier:  
Marketsource
CAS Number:  50-81-7
L(+)-Ascorbic acid 98%
CAS Number:  50-81-7
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
L-ASCORBIC ACID 25GM Catalog #TCA0537-025G

Product TCA0537-025G is similar to 103846-132 but manufacturer and specifications may vary.

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Caution: for Research Use Only

Synonyms:  Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
  • CAS Number:50-81-7
  • Formula:C₆H₈O₆
  • MDL Number:MFCD00064328
  • Molecular Weight:176.13 g/mol
  • Boiling Pt:552.7 °C (760 mm Hg)
  • Melting Pt:190…194 °C
  • Flash Pt:238.2 °C
  • Storage Temperature:Ambient

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.