Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

VWR® Screw Cap Microtubes with Socket Caps

Clear, 0.5 ml, Clear, Conical bottom, Insertable flat cap, No, 20000×g, Assembled, Sterile
Catalog # 76417-172
Supplier:   VWR International
VWR® Screw Cap Microtubes with Socket Caps
Catalog # 76417-172
Supplier:  VWR International
VWR® Screw Cap Microtubes with Socket Caps
VWR® Screw Cap Microtubes with Socket Caps

Clear, 0.5 ml, Clear, Conical bottom, Insertable flat cap, No, 20000×g, Assembled, Sterile

Price
$432.85
Log in for availability
Pack of 500 Case of 5000
Product Details & Documents

VWR® screw cap microtubes - Secure storage for valuable samples. VWR® screw cap microtubes provide ultra-pure polypropylene construction, leak-proof caps, and sterile/non sterile options for reliable lab sample storage.

  • Ultra-pure polypropylene (PP): Ensures chemical resistance and compliance with FDA 21 CFR 177.1520 & USP Class VI standards

  • High-clarity design: Crystal-clear walls for easy sample visibility
  • Secure O-ringed flat caps: ³/₄ turn design for leak-proof sealing and convenient opening/closing
  • Designed with an insertable ridge for convenient sample coding using colorful insert chips
  • Versatile temperature performance: Stable from −196 to +121 °C (not recommended for liquid nitrogen immersion)
  • Sterile and non sterile options: Choose pre-sterilized assemblies or convenient non-sterile combo packs
  • Multiple formats: Available in the three most commonly used volumes, with conical or skirted/free-standing bottoms for different applications

VWR® screw cap microtubes provide reliable, contamination-free storage for your critical samples, reagents, and biological materials. Designed for durability and clarity, these high-quality tubes are trusted in life science, clinical, and research laboratories worldwide. Ideal for sample storage, molecular biology, PCR, centrifugation, and general lab use.

 : Certified free of detectable DNase, RNase, DNA, and PCR inhibitors. Sterile items are also certified free of detectable endotoxins/pyrogens. Manufactured in compliance with stringent laboratory quality standards.

 : Spare caps and color insert chips also available.

Specifications
  • Cap type:
    Insertable flat cap
  • Format:
    Conical bottom
  • Sterility:
    Sterile
  • Volume:
    0.5 ml
  • Color:
    Clear
  • Packaging:
    Assembled
  • Max. rcf:
    20000×g
  • Cap color:
    Clear
  • Graduations:
    No