Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

γ-Cyclodextrine 98%

Glass bottle, 5 g
Catalog # 103835-932
Irritant
Marketsource
γ-Cyclodextrine 98%
Catalog # 103835-932
Supplier:  
Marketsource
CAS Number:  17465-86-0
γ-Cyclodextrine 98%

Glass bottle, 5 g

CAS Number:  17465-86-0
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
GAMMA-CYCLODEXTRIN 5GM Catalog #TCC0869-005G

 

Y-CYCLODEXTRIN 98% CRYSTAL 5G Catalog #76838-746

 

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

 : for Research Use Only

Synonyms:  gamma-Cyclodextrin, Cyclooctaamylose, γ-Cyclodextrin, Schardinger gamma-Dextrin
  • CAS Number:17465-86-0
  • Formula:C₄₈H₈₀O₄₀
  • MDL Number:MFCD00009595
  • Molecular Weight:1297.1248 g/mol
  • Melting Pt:267 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Size:
    5 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.