Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
5-Methoxyindole 99%

5-Methoxyindole 99%

Glass bottle, 5 g
Catalog # 103822-562
Irritant
Marketsource
5-Methoxyindole 99%
Catalog # 103822-562
Supplier:  
Marketsource
CAS Number:  1006-94-6
5-Methoxyindole 99%

Glass bottle, 5 g

CAS Number:  1006-94-6
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
5-METHOXYINDOLE MF=C9H9NO MW Catalog #101793-204

Product 101793-204 is similar to 103822-562 but may have a different manufacturer and/or packaging.

5-METHOXYINDOLE 99% 5G Catalog #AAAB25290-06

Product AAAB25290-06 is similar to 103822-562 but may have a different manufacturer and/or packaging.

5-METHOXYINDOLE 5G Catalog #CH01406-5G

Product CH01406-5G is similar to 103822-562 but may have a different manufacturer and/or packaging.

See more alternatives
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Caution: for Research Use Only

Synonyms:  5-Methoxy-1H-indole, Indol-5-yl methyl ether
  • CAS Number:1006-94-6
  • Formula:C₉H₉NO
  • MDL Number:MFCD00005674
  • Molecular Weight:147.18 g/mol
  • Boiling Pt:176…178 °C (760 mm Hg)
  • Melting Pt:52…55 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Size:
    5 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.