Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
CLASS 3.1 TEMP SAFETY DEVICE FOR FD 23
Catalog # 76410-386
Supplier:
Binder
Expand image
CLASS 3.1 TEMP SAFETY DEVICE FOR FD 23
Catalog # 76410-386
Supplier:
Binder
Supplier Number:
8012-1147