$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
CLASS3.3 INDEPENDENT TEMP DEVICE KBW240
76410-052
:
Binder
CLASS3.3 INDEPENDENT TEMP DEVICE KBW240
76410-052
:
Binder
:
8012-1750