Human Recombinant Complement Component C2 (from HEK293 Cells)
50 µg
Catalog # 103814-966
Marketsource
Supplier: ACRO Biosystems
Supplier #: CO2H52H750UG
Human Recombinant Complement Component C2 (from HEK293 Cells)
Catalog # 103814-966
Supplier: ACRO Biosystems
Marketsource
CAS Number:
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Human Complement Component C2 Protein, His Tag (MALS verified) (active enzyme), ACROBiosystems
Specifications
Specifications
- Size:50 µg
- Storage Conditions:–20 °C
- Molecular Weight:83.0 kDa
- Endotoxin Level:Low
- Purity:>90% as determined by SDS-PAGE.
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Source:HEK293 cells
- Cat. No.:103814-966
- Shipping Temperature:RT
- Protein/Peptide Type:Recombinant
- Gene ID:P06681-1
- Formulation:Lyophilized from 0.22 μm filtered solution in 20 mM Tris, 150 mM NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Protein Synonyms:Complement Component C2|Complement C2|C2
- Protein/Peptide Name:Complement Component C2
- Conjugation:Unconjugated
- Species:Human



