Human Recombinant Complement Component C2 (from HEK293 Cells)
1 mg
Catalog # 103814-964
Marketsource
Supplier: ACRO Biosystems
Supplier #: CO2H52H71MG
Human Recombinant Complement Component C2 (from HEK293 Cells)
Catalog # 103814-964
Supplier: ACRO Biosystems
Marketsource
CAS Number:
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Human Complement Component C2 Protein, His Tag (MALS verified) (active enzyme), ACROBiosystems
Specifications
Specifications
- Size:1 mg
- Storage Conditions:–20 °C
- Molecular Weight:83.0 kDa
- Endotoxin Level:Low
- Purity:>90% as determined by SDS-PAGE.
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Source:HEK293 cells
- Cat. No.:103814-964
- Shipping Temperature:RT
- Protein/Peptide Type:Recombinant
- Gene ID:P06681-1
- Formulation:Lyophilized from 0.22 μm filtered solution in 20 mM Tris, 150 mM NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Protein Synonyms:Complement Component C2|Complement C2|C2
- Protein/Peptide Name:Complement Component C2
- Conjugation:Unconjugated
- Species:Human
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



