Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Rapamycin ≥98%

50 mg
  103799-710
 
Marketsource
Rapamycin ≥98%
  103799-710
 :  
Marketsource
 :  53123-88-9
Rapamycin ≥98%

50 mg

 :  53123-88-9
 
 
 
RAPAMYCIN 99+% 50MG  AAJ62473-MF

Product AAJ62473-MF is similar to 103799-710 but manufacturer and specifications may vary.

 
   
 

Caution: Sealed in dry, store in freezer, under –20 °C.

 :  (3S,6R,7E,9R,10R,12R,14S,15E,17E,19E,21S,23S,26R,27R,34aS)-9,10,12,13,14,21,22,23,24,25,26,27,32,33,34,34a-Hexadecahydro-9,27-dihydroxy-3-[(1R)-2-[(1S,3R,4R)-4-hydroxy-3-methoxycyclohexyl]-1-methylethyl]-10,21-dimethoxy-6,8,12,14,20,26-hexamethyl-23,27-epoxy-3H-pyrido[2,1-c][1,4]oxaazacyclohentriacontine-1,5,11,28,29(4H,6H,31H)-pentone
  •  :53123-88-9
  •  :C₅₁H₇₉NO₁₃
  •  :MFCD00867594
  •  :914.19 g/mol
  •  :973 °C (760 mm Hg)
  •  :542.3 °C
  •  :Freezer
 

 

  • Size:
    50 mg

 

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.