GL-TIP GC 200UL 1 MG/200 UL 30 UG PACK96
Catalog # MSPP-782011201
Supplier: GL SCIENCES INC USA MS
GL-TIP GC 200UL 1 MG/200 UL 30 UG PACK96
Catalog # MSPP-782011201
Supplier: GL SCIENCES INC USA MS
Supplier Number: 7820-11201
Frequently Bought Together
Accessories for ThermoMixer® F
Supplier: Eppendorf
Dimethyl sulfoxide ≥99.9%, Ultra Pure Grade
Supplier: VWR
VWR® Disposable Serological Pipettes
Supplier: VWR International
Amersham ECL Western Blotting Select Detection Reagent, Cytiva
Supplier: Cytiva
Western blotting detection reagent, Amersham™ ECL™ Prime
Supplier: Cytiva
Disposable PD-10 Desalting Columns, Cytiva
Supplier: Cytiva
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Mix & Go!™ E. coli Transformation Kit and Buffer Set
Supplier: Zymo Research
Pepsin
Supplier: Promega Corporation
Blood Agar Plate, 5% Sheep Blood in Tryptic Soy Agar (TSA) Base, 15×100 mm plate
Supplier: Hardy Diagnostics
BUFFER TBS BLOCKING READY-TO-USE 500ML
Supplier: Li-Cor
Bis-Tris Mini Protein Gels, GenScript®
Supplier: Genscript
Detergent-OUT™ Tween® For Removal of Polysorbate Detergents, G-Biosciences
Supplier: G-Biosciences
COLUMN C18 INTEGRATD 25CMX75UM ID 1.7UM
Supplier: COANN TECHNOLOGIES MS
FlashGel™ DNA, RNA and Recovery Systems
Supplier: Lonza
Standard Metal Hub Luer Lock Needles
Supplier: Hamilton