Orders LinkContactUsLinkComponent
BioX Cryogenic Vials, Barcoded
BioX Cryogenic Vials, Barcoded
Barcoded cryovial, 1D/Human readable, 2.0 ml, Blue, Sterile
  76404-436
Supplier #:  BX88-6203
BioX Cryogenic Vials, Barcoded
  76404-436
BioX Cryogenic Vials, Barcoded
BioX Cryogenic Vials, Barcoded
Barcoded cryovial, 1D/Human readable, 2.0 ml, Blue, Sterile
$287.81
Case of 1,000
Barcoded polypropylene cryogenic vials with colored caps.
  • Suitable for liquid nitrogen storage
  • Pre-barcoded
  • O-ring free
  • Self standing
  • White writing patch and graduations
Unique permanent barcodes for superior traceability. 1D and human-readable on the side are perfect for hand scanning and the 2D barcodes on the bottom make these vials automation friendly.
Caution: Use in liquid phase LN₂ is not recommended.
  • Sterility:
    Sterile
  • Capacity:
    2.0 ml
  • Description:
    Barcoded cryovial, 1D/Human readable
  • Cap color:
    Blue