Orders LinkContactUsLinkComponent
BioX Cryogenic Vials, Barcoded
BioX Cryogenic Vials, Barcoded
Barcoded cryovial, 1D/Human readable, 0.5 ml, Blue, Sterile
Catalog # 76404-406
Supplier:   TWD Scientific
Supplier #:  BX88-6053
BioX Cryogenic Vials, Barcoded
Catalog # 76404-406
Supplier:  TWD Scientific
BioX Cryogenic Vials, Barcoded
BioX Cryogenic Vials, Barcoded
Barcoded cryovial, 1D/Human readable, 0.5 ml, Blue, Sterile
Price
$348.74
Log in for availability
Case of 1,000
Product Details & Documents
Barcoded polypropylene cryogenic vials with colored caps.
  • Suitable for liquid nitrogen storage
  • Pre-barcoded
  • O-ring free
  • Self standing
  • White writing patch and graduations
Unique permanent barcodes for superior traceability. 1D and human-readable on the side are perfect for hand scanning and the 2D barcodes on the bottom make these vials automation friendly.
Caution: Use in liquid phase LN₂ is not recommended.
Specifications
  • Sterility:
    Sterile
  • Capacity:
    0.5 ml
  • Description:
    Barcoded cryovial, 1D/Human readable
  • Cap color:
    Blue