TRIS-GLYCINE-SDS RUNNING BUFFER 10X 4L
Catalog # MSPP-BP-150
Supplier: BOSTON BIOPRODUCTS MS
TRIS-GLYCINE-SDS RUNNING BUFFER 10X 4L
Catalog # MSPP-BP-150
Supplier: BOSTON BIOPRODUCTS MS
Supplier Number: BP-150
Frequently Bought Together
Dulbecco's phosphate-buffered saline (DPBS), Mediatech
Supplier: Corning
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
VWR® Disposable Serological Pipettes
Supplier: VWR International
VWR® LTS Compatible Pipette Tips
Supplier: VWR International
Corning® PBS (Phosphate-Buffered Saline), Corning
Supplier: Corning
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® CryoPro® Fiberboard Storage Boxes and Dividers
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
Microcentrifuge Tube Lid Locks
Supplier: United Scientific Supplies
LENTI-X CONCENTRATOR
Supplier: Clontech Laboratories
ELISA KIT MOUSE INSULIN
Supplier: MERCODIA MS
BUFFER SDS-SAMPLE LAEMMLI 6X REDUCE 50ML
Supplier: BOSTON BIOPRODUCTS MS
Anti-LAT Mouse Monoclonal Antibody [clone: 11B.12]
Supplier: Biolegend
BUFFER TBST-WASH 10X W/1 TWEEN PH7.6 2L
Supplier: BOSTON BIOPRODUCTS MS
NUCLEOSPIN PLASMID EASYPURE
Supplier: Clontech Laboratories
Chameleon™ Duo Pre-stained Protein Ladder, LI-COR®
Supplier: Li-Cor